Skip to Content
MilliporeSigma
All Photos(1)

Documents

613570

Sigma-Aldrich

TIRAP Inhibitor Peptide, Cell-Permeable

The TIRAP Inhibitor Peptide, Cell-Permeable blocks LPS-, but not CpG-induced NF-κB activation. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

TIRAP Inhibitor Peptide, Cell-Permeable, Mal Peptide, MyD88-Adapter-Like Peptide, Toll-interleukin 1 Receptor (TIR) domain-containing Adapter Protein Peptide, Ant-Tirap 138-151, TIRAP Peptide, (RQIKIWFNRRMKWKKLQLRDAAPGGAIVS)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C163H268N52O38S
Molecular Weight:
3596.26
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable synthetic peptide containing mouse TIRAP138-151 fused to the Drosophila Antennapedia sequence. Specifically inhibits LPS-, but not CpG-induced NF-κB activation, PKR phosphorylation, and JNK phosphorylation in RAW.κB cells at ~40 µM. Also reported to block IκBα degradation.
A synthetic, cell-permeable peptide that corresponds to mouse toll-interleukin 1 receptor (TIR) domain-containing adapter protein 138-151 (TIRAP; also called Mal (MyD88-adapter-like), fused to the Drosophila antennapedia sequence. Shown to specifically inhibit LPS-induced, but not CpG-induced, NF-κB activation, PKR phosphorylation, and JNK phosphorylation in RAW.κB cells at ~40 µM. Also reported to block IκBα degradation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
LPS-induced NF-κB activation, PKR phosphorylation, and JNK phosphorylation in RAW.kB cells
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-Leu-Gln-Leu-Arg-Asp-Ala-Ala-Pro-Gly-Gly-Ala-Ile-Val-Ser-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Toshchakov, V., et al. 2002. Nat. Immunol.3, 392.
Fitzgerald, K.A., et al. 2001. Nature413, 78.
Henneke, P. and Golenbock, D.T. 2001. Nat. Immunol.2, 828.
Horng, T., et al. 2001. Nat. Immunol.2, 835.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service