Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6.09959

Supelco

Sodium hydroxide solution

c(NaOH) = 0.1 mol/l (0.1 N), concentrate for preparing 0.5 L volumetric solution for titration, Titriplex®

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352316

product name

Sodium hydroxide solution, for 500 ml, c(NaOH) = 0.1 mol/l (0.1 N) Titrisol

vapor pressure

3 mmHg ( 37 °C)

form

solution

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Mechanical and microstructural properties of iron mining tailings stabilized with alkali-activated binder produced from agro-industrial wastes.: This study demonstrates the use of sodium hydroxide solution to stabilize iron mining tailings with an alkali-activated binder derived from agro-industrial wastes. The focus is on enhancing mechanical properties and promoting sustainable waste management solutions (Levandoski WMK et al., 2023).


  • Improved liquid-liquid extraction followed by HPLC-UV for accurate and eco-friendly determination of tetramethylpyrazine in vinegar products.: Utilizing sodium hydroxide solution, this methodological study refines liquid-liquid extraction techniques for the determination of tetramethylpyrazine, highlighting advancements in analytical chemistry and ensuring the quality of vinegar products (Yang H et al., 2023).


  • Evaluation of ethanol-induced chitosan aerogels with human osteoblast cells.: Research uses sodium hydroxide solution to create chitosan aerogels, evaluating their biocompatibility and potential in regenerative medicine and bone tissue engineering, demonstrating their applicability in biomedical applications (Pantić M et al., 2023).


  • Synthesis and Biodegradation Test of a New Polyether Polyurethane Foam Produced from PEG 400, (L)-Lysine Ethyl Ester Diisocyanate (L-LDI) and Bis-hydroxymethyl Furan (BHMF).: This innovative study explores the synthesis and environmental impact of a new type of biodegradable polyurethane foam, utilizing sodium hydroxide solution in its production process, which is significant for sustainable material development (Olivito F et al., 2023).


  • A Mouse Model for Corneal Neovascularization by Alkali Burn.: The study details the use of sodium hydroxide solution to create a mouse model for corneal neovascularization, providing a valuable tool for ophthalmic research and potential treatments for eye diseases (Ammassam Veettil R et al., 2023).



Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service