Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

605206

Sigma-Aldrich

Thrombin, Citrate-Free, Human Plasma

Synonym(s):

Thrombin, Citrate-Free, Human Plasma

Sign Into View Organizational & Contract Pricing

Select a Size

100 U
$107.00

$107.00


Please contact Customer Service for Availability


Select a Size

Change View
100 U
$107.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

$107.00


Please contact Customer Service for Availability

biological source

human plasma

Quality Level

description

Merck USA index - 14, 9383

form

liquid

specific activity

≥2500 NIH units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Native thrombin from human plasma. Formulation useful for procedures in which the presence of citrate is undesirable.
Native thrombin from human plasma. Key enzyme in the coagulation cascade. Converts fibrinogen to fibrin and activates factor XIII, which cross-links and stabilizes the fibrin polymer. Formulation useful for procedures in which presence of citrate is undesirable.

Warning

Toxicity: Harmful (C)

Unit Definition

One unit is determined by comparison with a standard curve prepared using the Bureau of Biologics standard thrombin.

Physical form

In 100 mM NaCl, 20 mM Tris-HCl, pH 7.4.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Analysis Note

Homogeneous by SDS-PAGE

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service