Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

603425

Sigma-Aldrich

Threonine Dehydrogenase Inhibitor, QC1

The Threonine Dehydrogenase Inhibitor, QC1 controls the biological activity of Threonine Dehydrogenase.

Synonym(s):

Threonine Dehydrogenase Inhibitor, QC1, N-Benzyl-4-oxo-2-thioxo-3-(3-(trifluoromethyl)phenyl)-1,2,3,4-tetrahydroquinazoline-7-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H16F3N3O2S
CAS Number:
Molecular Weight:
455.45
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥96% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable quinazolinecarboxamide (QC) compound that acts as a potent, reversible and mixed noncompetitive inhibitor of threonine dehydrogenase activity (IC50 ~ 0.5 µM for mTDH) with excellent selectivity over hydroxysteroid, alcohol, lactate, or glucose-6-phosphate dehydrogenases (IC50 >10 µM). Shown to block threonine catabolism, induce autophagy and impede the growth of highly proliferating mouse embryonic stem cells (mESCs; EC50 ~3 µM) with no effect on ESCs-derived embryoid body cells, 3T3 fibroblasts or tumor cells (EC50 ~1 mM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Alexander, P.B., et al. 2011. Proc. Natl. Acad. Sci. USA108, 15828.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service