Skip to Content
MilliporeSigma
All Photos(1)

Documents

5980-OP

Millipore

Magnesium Chloride

OmniPur® Grade, complexometric: 99.0-102.0%

Synonym(s):

Magnesium chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.21

form

crystals

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Molecular Biology Grade.

Assay (complexometric): 99.0-102.0%
DNAse: none detected
Heavy metals (as Pb): ≤0.0005%
Insolubles: ≤.005%
Iron (Fe): ≤0.0005%
Nitrate (NO3): ≤0.001%
Phosphate (PO4): ≤0.0005%
Protease: none detected
RNAse: none detected
Sulfate (SO4): ≤0.002%





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Warning

Toxicity: Harmful (C)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service