Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

580559

Millipore

Paclitaxel, 6α-Hydroxy-

A metabolite of the anticancer agent Paclitaxel.

Synonym(s):

Paclitaxel, 6α-Hydroxy-, 6-Hydroxytaxol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H51NO15
CAS Number:
Molecular Weight:
869.91
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

colorless

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C47H51NO15/c1-24-30(61-43(57)33(51)32(27-16-10-7-11-17-27)48-41(55)28-18-12-8-13-19-28)22-47(58)40(62-42(56)29-20-14-9-15-21-29)36-45(6,38(54)35(60-25(2)49)31(24)44(47,4)5)37(53)34(52)39-46(36,23-59-39)63-26(3)50/h7-21,30,32-37,39-40,51-53,58H,22-23H2,1-6H3,(H,48,55)/t30-,32-,33+,34-,35+,36-,37-,39+,40-,45-,46+,47+/m0/s1

InChI key

NDCWHEDPSFRTDA-FJMWQILYSA-N

General description

A metabolite of the anticancer agent Paclitaxel (Cat. No. 580555).
A metabolite of the anticancer agent Paclitaxel (Cat. No. 580555). Note: 20 nmol = 17.4 µg.
Note: 20 nmol = 17.4 µg.

Biochem/physiol Actions

Cell permeable: no
Primary Target
A metabolite of the anticancer agent Paclitaxel
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Baldwin, S.J., et al. 1999. Br. J. Clin. Pharmacol.48, 424.
Fischer, V., et al. 1999. Drug Metab. Dispos.27, 410.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

953.8 - 1041.3 °F

Flash Point(C)

512.1 - 560.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service