Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

573109

Sigma-Aldrich

STAT3 Inhibitor VIII, 5,15-DPP

The STAT3 Inhibitor VIII, 5,15-DPP, also referenced under CAS 22112-89-6, controls the biological activity of STAT3.

Synonym(s):

STAT3 Inhibitor VIII, 5,15-DPP, 5,15-Diphenylporphyrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H22N4
CAS Number:
Molecular Weight:
462.54
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

deep blue to deep purple

solubility

DMSO: <1 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable porphyrin compound that binds STAT3 (KD = 880 nM by SPR) and prevents STAT3 SH2 domain-mediated ligand binding (IC50 = 280 nM) and dimerization (~60% inhibition at 50 µM in HEK293), while exhibiting much reduced activity against STAT1 ligand binding (IC50 = 10 µM) and no activity againt Grb2 ligand binding even at concentrations as high as 20 µM. One day 5,15-DPP pre-treatment (50 µM) of MDA-MB-468 cultures is shown to greatly inhibit IL-6-induced STAT3, but not STAT1/5a/5b, nuclear translocation (by >90%) and nuclear STAT3 c-myc promoter binding activity, resulting in a more than 90% reduction in cellular c-myc protein level.
A cell-permeable porphyrin compound that directly binds to STAT3, prevents STAT3 dimerization and acts as a STAT3-SH2 domain antagonist with selectivity over STAT1 and Grb2 (IC50 = 0.28, 10 and >20 µM, respectively). Shown to preferentially suppress STAT3 nuclear translocation and subsequent DNA binding in MDA-MB-468 cells with negligible effect on STAT1, STAT5a and STAT5b; further, reduce c-myc promoter binding and c-myc protein expression in IL-6 induced HEK-293-STAT3 cells. Does neither affect JAK kinase activity nor lower the expression or the phosphorylation levels of STAT3.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Uehara, Y., et al. 2009. Biochem. Biophys. Res. Commun.380, 627.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service