Skip to Content
MilliporeSigma
All Photos(1)

Documents

572888-M

Millipore

SU1498

A potent, reversible, ATP-competitive and selective inhibitor of Flk-1 kinase (IC₅₀ = 700 nM), a vascular endothelial growth factor (VEGF) receptor kinase.

Synonym(s):

SU1498, (E)-3-(3,5-Diisopropyl-4-hydroxyphenyl)-2-[(3-phenyl-n-propyl)amino-carbonyl]acrylonitrile, VEGFR Tyrosine Kinase Inhibitor VIII, VEGFR2 Kinase Inhibitor X

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N2O2
Molecular Weight:
390.52
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

potency

700 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 5 mg/mL

storage temp.

−20°C

General description

A potent, reversible, ATP-competitive and selective inhibitor of Flk-1 kinase (IC50 = 700 nM), a vascular endothelial growth factor (VEGF) receptor kinase. Also reduces the expression of ets-1, a transcription factor stimulated by VEGF. Has only a weak inhibitory effect on PDGF-receptor (IC50 >50 µM), EGF-receptor (IC50 >100 µM), and HER2 (IC50 >100 µM) kinases. Acts as an angiogenesis inhibitor as shown by its activity in the chorioallantoic membrane (CAM) assay and in an in vivo VEGF-induced permeability assay.
A potent, reversible, ATP-competitive and selective inhibitor of Flk-1 kinase (IC50 = 700 nM), a vascular endothelial growth factor (VEGF) receptor kinase. Has only a weak inhibitory effect on PDGF receptor (IC50 >50 µM), EGF-receptor (IC50 >100 µM), and HER-2 (IC50 >100 µM) kinases. Acts as an angiogenesis inhibitor as shown by its activity in the chorioallantoic membrane (CAM) assay and in an in vivo VEGF-induced permeability assay.

Biochem/physiol Actions

Primary Target
Flk-1 kinase

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Arbiser, J.L., et al. 2000. Am. J. Pathol. 156, 1469.
Strawn, L.M., et al. 1996. Cancer Res. 56, 3540.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service