Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

572650

Sigma-Aldrich

SU9516

A cell-permeable 3-substituted indolinone compound that displays anti-proliferative and proapoptotic properties in tumor cells.

Synonym(s):

SU9516, 3-[1-(3H-Imidazol-4-yl)-meth-(Z)-ylidene]-5-methoxy-1,3-dihydro-indol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N3O2
CAS Number:
Molecular Weight:
241.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow to orange

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[nH]1cncc1C=C2c3c(ccc(c3)OC)NC2=O

InChI

1S/C13H11N3O2/c1-18-9-2-3-12-10(5-9)11(13(17)16-12)4-8-6-14-7-15-8/h2-7H,1H3,(H,14,15)(H,16,17)

General description

A cell-permeable 3-substituted indolinone compound that displays anti-proliferative and proapoptotic properties in tumor cells. Acts as a potent, selective, reversible, and ATP-competitive inhibitor of cyclin dependent kinases (Cdks; IC50 = 22 nM for Cdk2/A; 40 nM for Cdk1/B; 200 nM for Cdk4/D1). It shows no significant effect (IC50 >10 µM) on the activities of PKC, p38, PDGFRβ, or EGFR. Inhibits the proliferation of growth factor-stimulated colon carcinoma cells by binding to Cdk2 and preventing the phosphorylation of pRb and its dissociation from E2F.
A cell-permeable, 3-substituted indolinone compound that displays anti-proliferative and pro-apoptotic properties in tumor cells. Acts as a potent, selective, reversible, and ATP-competitive inhibitor of cyclin dependent kinases (Cdks) (IC50 = 22 nM for Cdk2/A; 40 nM for Cdk1/B; 200 nM for Cdk4/D1). Exhibits no significant effect on the activities of PKC, p38, PDGFRβ, or EGFR (IC50 >10 µM). Reported to inhibit the proliferation of growth factor-stimulated colon carcinoma cells by binding to Cdk2 and preventing the phosphorylation of pRb and its dissociation from E2F.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cdk2/A
Product competes with ATP.
Reversible: yes
Target IC50: 22 nM for Cdk2/A; 40 nM for Cdk1/B; 200 nM for Cdk4/D1

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Yu, B., et al. 2002. Biochem. Pharmacol.64, 1091.
Lane, M.E., et al. 2001. Cancer Res.61, 6170.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service