Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

567806

Sigma-Aldrich

Src Kinase Inhibitor II

The Src Kinase Inhibitor II, also referenced under CAS 459848-35-2, controls the biological activity of Src Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Src Kinase Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N4O3S
CAS Number:
Molecular Weight:
420.48
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale orange

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A potent, selective, reversible, and ATP-competitive inhibitor of Src family tyrosine kinases (IC50 = 1.2 µM for human recombinant Csk). Exhibits little effect against p38 MAPK or FGFR tyrosine kinase even at concentrations as high as 50 µM.
A substituted triazoloquinoline-1-thione compound that acts as a potent, ATP-competitive, reversible and selective inhibitor of Src family tyrosine kinases (IC50 = 1.2 µM for human recombinant Csk). Exhibits little effect against p38 MAPK or FGFR even at concentrations as high as 50 µM.

Biochem/physiol Actions

Cell permeable: no
Primary Target
human recombinant Csk
Product competes with ATP.
Reversible: yes
Target IC50: 1.2 µM for human recombinant Csk

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kilimnik, A., et al. 2003. Cell. Mol. Biol. Lett.8, 588.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service