Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

567693

Sigma-Aldrich

SOST, Mouse, Recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Assay

≥90% (SDS-PAGE)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, mouse sclerostin (SOST) consisting of amino acids 24-211, expressed in NS0-derived murine myeloma cells with an N-terminal His•Tag. SOST is expressed by osteoclasts in developing bones of mouse embryos and acts as a negative regulator of bone formation through repression of BMP-induced osteogenesis.

Biochem/physiol Actions

Inhibition Assay: Inhibits hBMP-6-induced alkaline phosphatase by 50-80% in MC3T3-E1 mouse osteoblastic cells when used at a concentration of 30 µg/ml.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm filter-sterilized PBS, BSA.

Reconstitution

Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 200 µg/ml in sterile PBS containing at least 0.1% human or bovine serum albumin.

Other Notes

Kusu, N. et al. 2003. J. Biol. Chem.278, 24113.
Balemans, W. et al. 2001. Hum. Mol. Genet.10 537.
Brunkow, M.E. et al. 2001. Am. J. Hum. Genet.68, 577.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachel C Nordberg et al.
PloS one, 14(10), e0223245-e0223245 (2019-10-05)
Mechanical loading is essential for the maintenance of musculoskeletal homeostasis. Cartilage has been demonstrated to be highly mechanoresponsive, but the mechanisms by which chondrocytes respond to mechanical stimuli are not clearly understood. The goal of the study was to determine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service