Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56766-M

Sigma-Aldrich

Spermidine, Trihydrochloride

Polyamine that plays an important role in the regulation of cellular proliferation and differentiation.

Synonym(s):

Spermidine, Trihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$315.00

$315.00


Please contact Customer Service for Availability


Select a Size

Change View
5 G
$315.00

About This Item

Empirical Formula (Hill Notation):
C7H19N3 · 3HCl
CAS Number:
Molecular Weight:
254.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$315.00


Please contact Customer Service for Availability

description

Merck USA index - 14, 8742

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

ethanol: soluble
water: soluble

storage temp.

10-30°C

SMILES string

[Cl-].[Cl-].[Cl-].[N+H2](CCC[N+H3])CCCC[N+H3]

InChI

1S/C7H19N3.3ClH/c8-4-1-2-6-10-7-3-5-9;;;/h10H,1-9H2;3*1H

InChI key

LCNBIHVSOPXFMR-UHFFFAOYSA-N

General description

Polyamine that plays an important role in the regulation of cellular proliferation and differentiation. Inhibits neuronal nitric oxide synthase.

Biochem/physiol Actions

Primary Target
Inhibits neuronal nitric oxide synthase

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C).

Other Notes

Hu, J., et al. 1994. Neurosci. Lett. 175, 41.
Trubiani, O., et al. 1994. Biochem. Mol. Biol. Int.32, 1085.
He, Y., et al. 1993. Eur. J. Biochem. 217, 89.
Svensson, F., et al. 1993. Mol. Cell. Biochem.124, 141.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service