Skip to Content
MilliporeSigma
All Photos(1)

Documents

567422

Millipore

Sodium Acetate, 3M, pH 5.2, Molecular Biology Grade

Sterile solution. Prepared with DEPC for RNA applications. Also suitable for DNA applications.

Synonym(s):

Sodium Acetate, 3M, pH 5.2, Molecular Biology Grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3O2 · Na
CAS Number:
Molecular Weight:
82.03
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.25

grade

Molecular Biology

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

foreign activity

DNases, RNases, none detected

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Related Categories

General description

Sterile solution. Prepared with DEPC for RNA applications. Also suitable for DNA applications.
Supplied as a 3 M aqueous solution, treated with DEPC for use in RNA applicatons. Also suitable for DNA applications. Sodium acetate is widely used in molecular biology applications as a buffer and in the purification and precipitation of nucleic acids, protein crystallization, staining of protein gels, and HPLC.

Warning

Toxicity: Harmful (C)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service