Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

565784-M

Millipore

γ-Secretase Inhibitor IX

≥95% (HPLC), liquid, γ-secretase inhibitor, Calbiochem®

Synonym(s):

InSolution γ-Secretase Inhibitor IX, N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, DAPT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26F2N2O4
Molecular Weight:
432.46
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Product Name

γ-Secretase Inhibitor IX, InSolution, ≥95%, 25 mM, g-secretase Inhibitor IX

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

clear colorless

storage temp.

2-8°C

General description

A cell-permeable dipeptide that inhibits γ-secretase activity and suppresses Aβ production (Aβ total IC50 = 115 nM; Aβ42 IC50 = 200 nM). Reported to be functionally active in both HEK293 cells overexpressing human APP751 and neuronal cultures, however, it does not affect the secretion of amyloid-b precursor protein (APP). Effective in acutely lowering Ab in APPV717F-transgenic mice. Also effective in inhibiting the production of a ~6 kDa C-terminal cleavage product (CTFg) in HEK293 cells stably transfected with the Swedish mutant, βAPP695. Reported to reduce extracellular Aβ plaques and intracellular Aβ accumulation in 3xTgAD transgenic mice.

Biochem/physiol Actions

Primary Target
total
Target IC50: 115 nM, 200 nM against Aβtotal, Aβ42, respectively

Warning

Toxicity: Irritant (B)

Physical form

A 25 mM (5 mg/462 µl or 10 mg/924 µl) solution of γ-Secretase Inhibitor IX (Cat. No. 565770) in DMSO.

Reconstitution

Following initial use, aliquot and freeze (-20°C).

Other Notes

Oddo, S., et al. 2004. Neuron43, 321.
Dovey, H.F., et al. 2001. J. Neurochem.76, 173.
Vandermeeren, M., et al. 2001. Neurosci. Lett.315, 145.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (refers to pure substance)

Flash Point(C)

87 °C - closed cup - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service