Skip to Content
MilliporeSigma
All Photos(1)

Documents

565780

Sigma-Aldrich

β-Secretase Inhibitor III

The β-Secretase Inhibitor III controls the biological activity of β-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

β-Secretase Inhibitor III, Inhibitor GL189, H-EVNstatineVAEF-NH₂

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H70N10O14
Molecular Weight:
963.09
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

DMSO: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A substrate analog inhibitor of β-secretase (BACE) that completely blocks proteolytic activity in solubilized membrane fractions of BACE-transfected MDCK cells at a concentration of 5 µM.
A substrate analog inhibitor of β-secretase (BACE) that completely blocks the proteolytic activity (at 5 µM) in solubilized membrane fractions from BACE transfected MDCK cells.

Biochem/physiol Actions

Cell permeable: no
Effective concentration: 5 µM completely blocking the proteolytic activity of β-secretase (BACE) in solubilized membrane fractions from BACE transfected MDCK cells
Primary Target
BACE
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Glu-Val-Asn-Statine-Val-Ala-Glu-Phe-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Capell, A., et al. 2002. J. Biol. Chem.277, 5637.
Tung, J.S., et al. 2002. J. Med. Chem.45, 259.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inmaculada Lopez-Font et al.
Molecular neurobiology, 56(12), 8603-8616 (2019-07-11)
The β-site amyloid precursor protein cleaving enzyme 1 (BACE1) is the main brain β-secretase responsible for the amyloidogenic processing of the amyloid precursor protein (APP). Previous studies have suggested that cerebrospinal fluid (CSF) β-secretase activity may be a candidate diagnostic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service