Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

565755

Sigma-Aldrich

γ-Secretase Inhibitor II

The γ-Secretase Inhibitor II controls the biological activity of γ-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

γ-Secretase Inhibitor II, MW167

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$373.00

$373.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
$373.00

About This Item

Empirical Formula (Hill Notation):
C33H57F2N5O9
Molecular Weight:
705.83
UNSPSC Code:
12352200
NACRES:
NA.54

$373.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 10 mM

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, reversible and selective peptidomimetic inhibitor of γ-secretase with an IC50 of 13 µM for the inhibition of amyloid β-protein (Aβ) production in β-amyloid precursor protein (APP)-transfected Chinese hamster ovary cells. Displays only weak inhibitory activity against calpain II (IC50 = 100 µM in a purified enzyme assay).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
γ-secretase
Product does not compete with ATP.
Reversible: yes
Target IC50: 13 µM against γ-secretase

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wolfe, M.S., et al. 1999. Biochemistry38, 4720.
Wolfe, M.S., et al. 1998. J. Med. Chem. 41, 6.
Citron, M., et al. 1996. Proc. Natl. Acad. Sci. USA93, 13170.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaxiong Yang et al.
Journal of neurochemistry, 88(6), 1361-1372 (2004-03-11)
Glutamate uptake by high affinity glutamate transporters is essential for preventing excitotoxicity and maintaining normal synaptic function. We have discovered a novel role for presenilin-1 (PS1) as a regulator of glutamate transport. PS1-deficient neurons showed a decrease in glutamate uptake

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service