Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

559393-M

Millipore

SB 202190, Hydrochloride

A water-soluble form of the potent p38 MAP kinase inhibitor SB 202190.

Synonym(s):

SB 202190, Hydrochloride, 4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)1H-imidazole, HCl, p38 MAP Kinase Inhibitor II, Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14FN3O · xHCl
CAS Number:
Molecular Weight:
331.34 (free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

potency

16 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

amber

solubility

water: 0.5 mg/mL (slightly soluble)
DMSO: soluble

storage temp.

−20°C

SMILES string

Fc1ccc(cc1)c2[nH]c(nc2c4ccncc4)c3ccc(cc3)O.Cl

InChI

1S/C20H14FN3O.ClH/c21-16-5-1-13(2-6-16)18-19(14-9-11-22-12-10-14)24-20(23-18)15-3-7-17(25)8-4-15;/h1-12,25H,(H,23,24);1H

InChI key

XOQSUTIMRLPFPB-UHFFFAOYSA-N

General description

A water-soluble form of the potent p38 MAP kinase inhibitor SB 202190 (Cat. No. 559388).
A water-soluble form of the potent p38 MAP kinase inhibitor SB 202190 (Cat. No. 559388).

Biochem/physiol Actions

Target IC50: 350 nM against the kinase activity of p38β; 280 nM against p38 phosphorylation of activating transcription factor 2 (ATF-2)

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 weeks at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service