Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

559292

Sigma-Aldrich

RSK Inhibitor, SL0101

InSolution, ≥95%

Synonym(s):

InSolution RSK Inhibitor, SL0101

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24O12
Molecular Weight:
516.45
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable Kaempferol (Cat. No. 420345) glycoside that targets the the N-terminal kinase domain of p90 ribosomal S6 kinase and inhibits RSK kinase activity (IC50 = 89 nM with 10 µM ATP) in a selective, reversible, and ATP-competitive (Ki = 1 µM) manner. Shown to inhibit the proliferation of MCF-7, but not the normal breast cell line MCF-10A, and specifically block PDB-induced cellular phosphorylation of RSK substrate p140, but not that of RSK or RSK upstream kinases even at concentrations as high as 100 µM. The solid form (Cat. No. 559285) is also available.
A cell-permeable Kaempferol (Cat. No. 420345) glycoside that targets the the N-terminal kinase domain of p90 ribosomal S6 kinase and inhibits RSK kinase activity (IC50 = 89 nM with 10 µM ATP) in a selective, reversible, and ATP-competitive (Ki = 1 µM) manner. Shown to inhibit the proliferation of MCF-7, but not the normal breast cell line MCF-10A, and specifically block PDB-induced cellular phosphorylation of RSK substrate p140, but not that of RSK or RSK upstream kinases even at concentrations as high as 100 µM.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (500 µg/97 µl) solution of RSK Inhibitor, SL0101 (Cat. No. 559285) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Smith, J.A., et al. 2007. Bioorg. Med. Chem.15, 5018.
Smith, J.A., et al. 2006. Bioorg. Med. Chem.14, 6043.
Xu, Y.M., et al. 2006. Bioorg. Med. Chem.14, 3974.
Maloney, D.J. and Hecht, S.M., 2005. Org. Lett.7, 1097.
Smith, J.A., et al. 2005. Cancer Res.65, 1027.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service