Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

557404-M

Millipore

RNA Polymerase III Inhibitor

InSolution, ≥97%

Synonym(s):

InSolution RNA Polymerase III Inhibitor, 2-Chloro-N-(3-(5-chloro-3-methylbenzo[b]thiophen-2-yl)-1-methyl-1H-pyrazol-5-yl)benzenesulfonamide, N-(1-(3-(5-Chloro-3-methylbenzo[b]thiophen-2-yl-1-methyl-1H-pyrazol-5-yl))-2-chlorobenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15Cl2N3O2S2
CAS Number:
Molecular Weight:
452.38
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

liquid

potency

27 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

SMILES string

[S](=O)(=O)(Nc2[n](nc(c2)c3[s]c4c(c3C)cc(cc4)Cl)C)c1c(cccc1)Cl

InChI

1S/C19H15Cl2N3O2S2/c1-11-13-9-12(20)7-8-16(13)27-19(11)15-10-18(24(2)22-15)23-28(25,26)17-6-4-3-5-14(17)21/h3-10,23H,1-2H3

InChI key

BVBDTTLISMIOJY-UHFFFAOYSA-N

General description

A cell-permeable inhibitor of RNA polymerase III (RNA Pol III) (IC50 = 27 µM for human RNA Pol III and 32 µM for Saccharomyces cerevisiae RNA Pol III). Inhibits cell growth in yeast by preventing RNA Pol III-mediated tRNA transcription. The solid form of this compound (Cat. No. 557403) is also available.

Biochem/physiol Actions

Primary Target
RNA Polymerase III

Warning

Toxicity: Standard Handling (A)

Physical form

A 25 mM (5 mg/442 µL) solution of RNA Polymerase III Inhibitor (Cat. No. 557403) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -70°C.

Other Notes

Wu, L., et al. 2003. Euk. Cell2, 256.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (refers to pure substance)

Flash Point(C)

87 °C - closed cup - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service