Skip to Content
MilliporeSigma
All Photos(1)

Documents

555555

Sigma-Aldrich

Rho Kinase Inhibitor V

The Rho Kinase Inhibitor V, also referenced under CAS 1072906-02-5, controls the biological activity of Rho Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Rho Kinase Inhibitor V, N-(4-(1H-pyrazol-4-yl)phenyl)-2,3-dihydrobenzo[b][1,4]dioxine-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15N3O3
CAS Number:
Molecular Weight:
321.33
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

ethanol: 2 mg/mL
DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C18H15N3O3/c22-18(17-11-23-15-3-1-2-4-16(15)24-17)21-14-7-5-12(6-8-14)13-9-19-20-10-13/h1-10,17H,11H2,(H,19,20)(H,21,22)

InChI key

MCQBNMPALTYSES-UHFFFAOYSA-N

General description

A cell-permeable pyrazolyl carboxamide compound that acts as a potent, ATP-binding pocket-targeting inhibitor of ROCK-II (IC50 = 1.5 nM) and effectively inhibits the myosin light chain 2 phosphorylation on Thr18/Ser19 in A7r5 cells (IC50 = 72 nM). It afftects the activities of PKA, MRCK, Akt1 (IC50 = 0.186, 1.19, and 1.41 µM, respectively) and CYP-450 enzymes (% inhibition of 2C9/2D6/3A4/1A2 at 10 µM = 87/40/1/20, respectively) only at much higher concentrations.
Feng, Y., et al. 2008. J. Med. Chem.51, 6642.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service