Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.55364

Millipore

DiluCult Dispensing Tubing

DiluCult & DiluCult 2 accessory

Synonym(s):

Dispensing Hose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.84
Pricing and availability is not currently available.

manufacturer/tradename

DiluCult

application(s)

food and beverages
sample preparation

storage temp.

2-30°C

General description

DiluCult & DiluCult 2

Legal Information

DILUCULT is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DiluCult™ gravimetric dilutors automate food sample dilution, enhancing workflow efficiency in sample preparation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service