Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

553514-M

Millipore

RAD51 Inhibitor II, RI-1

The RAD51 Inhibitor, RI-1 controls the biological activity of RAD51. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

RAD51 Inhibitor II, RI-1, 3-chloro-1-(3,4-dichlorophenyl)-4-morpholino-1H-pyrrole-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11Cl3N2O3
Molecular Weight:
361.61
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (HPLC)

form

solid

potency

5-30 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

bright yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable 1H-Pyrrole-2,3-dione derivative that covalently binds to RAD51 at Cys319 and irreversibly destabilizes a protein-protein interface that is essential for filament formation and recombinase activity. Stimulates single strand annealing at a 5-30 µM concentration. Inhibits subnuclear RAD51 foci formation following DNA damage without affecting replication protein A. Also, effectively sensitizes cancer cells to mitomycin C (~25 µM) and generate synergistic cell kill in RAD51C mutant cells and with lesser effect on RAD51 complemented cells. Satisfies all four Lipinski rules for predicting drug likeness.

Biochem/physiol Actions

Primary Target
RAD51

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Budke, B., et al. 2012. Nucleic Acids Res.40, 7347.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service