Skip to Content
MilliporeSigma
All Photos(1)

Documents

553513

Sigma-Aldrich

Rac Inhibitor III

EHop-016

Synonym(s):

EHop-016, N4-(9-Ethyl-9H-carbazol-3-yl)-N2-[3-(4-morpholinyl)propyl]-2,4-pyrimidinediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N6O
CAS Number:
Molecular Weight:
430.55
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

SMILES string

CCn1c2ccccc2c3cc(Nc4ccnc(NCCCN5CCOCC5)n4)ccc13

InChI

1S/C25H30N6O/c1-2-31-22-7-4-3-6-20(22)21-18-19(8-9-23(21)31)28-24-10-12-27-25(29-24)26-11-5-13-30-14-16-32-17-15-30/h3-4,6-10,12,18H,2,5,11,13-17H2,1H3,(H2,26,27,28,29)

InChI key

AFTZZRFCMOAFCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

EHop-016 is a potent and specific inhibitor of Rac1 and Rac3 GTPase activity that inhibits Vav2 interaction with Rac, Rac-activated PAK1, lamellipodia formation, and cell migration. EHop-016 inhibits the Rac activity of MDA-MB-231 metastatic breast cancer cells and reduces Rac-directed lamellipodia formation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service