Skip to Content
MilliporeSigma
All Photos(1)

Documents

553504

Sigma-Aldrich

RAD51, His•Tag®, Pneumocystis carinii, Recombinant, E. coli

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.42

Assay

≥95% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, Pneumocystis carinii RAD51 fused at the N-terminus to a His•Tag sequence and expressed in E. coli. The Pneumocystis carinii RAD51 is equivalent to bacterial RecA and the homologue of yeast RAD51. It plays a pivotal role in the homologous recombination pathway for DNA double-strand break (DSB) repair. RAD51 has been shown to colocalize with tumor suppressor proteins BRCA1 (Cat. No. 203659) and BRCA2 in DNA-damage-induced foci and RAD51 is known to interact directly with BRCA2.
Recombinant, Pneumocystis carinii RAD51 fused at the N-terminus to a His•Tag sequence and expressed in E. coli. The Pneumocystis cariniiRAD51 is equivalent to bacterial RecA and the homolog of yeast RAD51. It plays a pivotal role in the homologous recombination pathway for DNA double-strand break (DSB) repair. RAD51 has been shown to colocalize with tumor suppressor proteins BRCA1 (Cat. No. 203659) and BRCA2 in DNA-damage-induced foci and RAD51 is known to interact directly with BRCA2.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit equals 1 ng of purified protein. 20 units are sufficient for a DNA-protein assay and 100 units are sufficient for a protein-protein interaction assay.

Physical form

In 100 mM KCl, 20 mM Tris-HCl, 1 mM DTT, 0.2 mM EDTA, 20% glycerol, pH 8.0.

Other Notes

West, S.C. 2003. Nat. Rev. Mol. Cell Biol.4, 435.
Jasin, M. 2002. Oncogene21, 8981.
Symington, L.S., et al. 2002. Microbiol. Mol. Biol. Rev.66, 630.
Daboussi, F., et al. 2002. Cell. Signal.14, 969.
Liu, Y. and West, S.C. 2002. Breast Cancer Res.4, 9.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service