Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

553325

Sigma-Aldrich

Ras, Human, Recombinant, Wild-Type, E. coli

Synonym(s):

rH-Ras, WT

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$550.00

$550.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$550.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$550.00


Please contact Customer Service for Availability

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human wild-type Ras expressed in E. coli. Suitable as a substrate for farnesyltransferase.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 45 mM NaCl, 20 mM Tris-HCl, 5 mM MgCl₂, 1 mM DTT, 50% glycerol, pH 8.0.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Marshall, M.S. 1995. FASEB J.9, 1311.
James, G.L., et al. 1993. Science260, 1937.
Reiss, Y., et al. 1990. Cell62, 81.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service