Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

553014

Sigma-Aldrich

Raf Kinase Inhibitor IV

The Raf Kinase Inhibitor IV, also referenced under CAS 303727-31-3, controls the biological activity of Raf Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Raf Kinase Inhibitor IV, 2-(Phenyl)-4-(3-hydroxy-4-chlorophenyl)-5-(4-pyridyl)-1H-imidazole, L 779450

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14ClN3O
CAS Number:
Molecular Weight:
347.80
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥97% (HPLC)

form

solid

potency

10 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

ethanol: 10 mg/mL
DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1c(cc(cc1)c2nc([nH]c2c4ccncc4)c3ccccc3)O

InChI

1S/C20H14ClN3O/c21-16-7-6-15(12-17(16)25)19-18(13-8-10-22-11-9-13)23-20(24-19)14-4-2-1-3-5-14/h1-12,25H,(H,23,24)

InChI key

WXJLXRNWMLWVFB-UHFFFAOYSA-N

General description

A cell-permeable triarylimidazole compound that acts as a reversible, ATP-competitive, and highly potent inhibitor of Raf kinase (IC50 = 10 nM & Kd = 2.4 nM for B-raf) with 7-, 30-, and 70-fold selectivity over p38α, GSK-3β, and Lck, respectively. Cellular data have indicated that the compound is more effective towards Raf-1and A-Raf than B-Raf.
A cell-permeable triarylimidazole compound that acts as a reversible, ATP-competitive, and highly potent inhibitor of Raf kinase (IC50 = 10 nM and Kd = 2.4 nM for B-raf) with 7-, 30-, and 70-fold selectivity over p38α, GSK-3β, and Lck, respectively. Cellular data have indicated that the compound is more effective towards Raf-1and A-Raf than B-Raf.

Biochem/physiol Actions

Cell permeable: yes
Kd = 2.4 nM for B-raf
Primary Target
Raf Kinase
Product competes with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Other Notes

Takle, A.K., et al. 2006. Bioorg. Med. Chem. Lett.16, 378.
Shelton, J.G., et al. 2003. Leukemia17, 1765.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service