Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

540218-M

Sigma-Aldrich

LYP Inhibitor II, LTV-1

The LYP Inhibitor II, LTV-1 controls the biological activity of LYP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

LYP Inhibitor II, LTV-1, ( E)-3-((4-((4,6-Dioxo-2-thioxo-1- o-tolyltetrahydropyrimidin-5(6H)-ylidene)methyl)phenoxy)methyl)benzoic acid, Lymphoid Tyrosine Phosphatase Inhibitor II, LTV-1, (E)-3-((4-((4,6-Dioxo-2-thioxo-1-o-tolyltetrahydropyrimidin-5(6H)-ylidene)methyl)phenoxy)methyl)benzoic acid, Lymphoid Tyrosine Phosphatase Inhibitor II, LTV-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H20N2O5S
Molecular Weight:
472.51
UNSPSC Code:
12352200
NACRES:
NA.28
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable thiobarbituryl-benzoate compound that acts as a highly potent and reversible inhibitor of lymphoid tyrosine phosphatase (LYP) (IC50 = 508 nM) and thereby enhances TCR signaling in intact cells. Exhibits competitive to mixed mode of inhibition (Ki = 384 nM). Also shown to inhibit the mutant, disease associated LYP-Trp620 activity. Binds to the phosphate-binding loop and mimicks the phosphotyrosine moiety of substrates. Displays acceptable selectivity over a wide range of protein phosphatases (TCPTP, PTP1B, PEP (mouse LYP), SHP1, CD45 and PTP-PEST: IC50 = 1.52, 1.59, 7.56, 23.2, 30.1, and 100 µM, respectively). Does not exhibit any toxicity at pharmacological doses when tested in primary human cell lines.

Biochem/physiol Actions

Primary Target
LYP
Secondary Target
Othet PTPases
Target IC50: 508 nM for LYP

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C>

Other Notes

Vang, T., et al. 2012. Nat. Chem. Biol.8, 437.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service