Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

540217

Sigma-Aldrich

PTP LYP Inhibitor

The PTP LYP Inhibitor controls the biological activity of PTP LYP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PTP LYP Inhibitor, (2-pyridine)(Ph)₂P-Au-Cl, HePTP Inhibitor III, PTP Inhibitor XXVIII, PTP-PEST Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14AuClNP
Molecular Weight:
495.69
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, Au(I)-phosphine complex that acts as selective inhibitor of lymphoid tyrosine phosphatase (LYP) (IC50= 1.5 µM) and has a ~10-fold selectivity for LYP over PTP-PEST, HePTP, and CD45 in both JTAg cells and primary mouse thymocytes.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Karver, M.R., et al. 2009. J. Med. Chem.52, 6912.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service