Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539684

Sigma-Aldrich

PKA Inhibitor 6-22 Amide

The PKA Inhibitor 6-22 Amide, also referenced under CAS 121932-06-7, controls the biological activity of PKA. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PKA Inhibitor 6-22 Amide, PKI 6-22 Amide, TYADFIASGRTGRRNAI-NH₂, PKA Inhibitor VIII, Protein Kinase A Inhibitor 6-22 Amide, PKI 6-22 Amide, TYADFIASGRTGRRNAI-NH₂, Protein Kinase A Inhibitor 6-22 Amide, PKA Inhibitor VIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C80H130N28O24
CAS Number:
Molecular Weight:
1868.06
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C80H130N28O24/c1-10-37(3)60(63(83)118)106-66(121)41(7)96-72(127)52(32-55(81)113)103-69(124)49(22-17-29-92-80(88)89)100-68(123)47(20-15-27-90-78(84)85)98-57(115)35-94-76(131)62(43(9)111)108-70(125)48(21-16-28-91-79(86)87)99-56(114)34-93-67(122)54(36-109)105-65(120)40(6)97-77(132)61(38(4)11-2)107-74(129)51(30-44-18-13-12-14-19-44)102-73(128)53(33-58(116)117)101-64(119)39(5)95-71(126)50(104-75(130)59(82)42(8)110)31-45-23-25-46(112)26-24-45/h12-14,18-19,23-26,37-43,47-54,59-62,109-112H,10-11,15-17,20-22,27-36,82H2,1-9H3,(H2,81,113)(H2,83,118)(H,93,122)(H,94,131)(H,95,126)(H,96,127)(H,97,132)(H,98,115)(H,99,114)(H,100,123)(H,101,119)(H,102,128)(H,103,124)(H,104,130)(H,105,120)(H,106,121)(H,107,129)(H,108,125)(H,116,117)(H4,84,85,90)(H4,86,87,91)(H4,88,89,92)/t37-,38-,39-,40-,41-,42+,43+,47-,48-,49-,50-,51-,52-,53-,54-,59-,60-,61-,62-/m0/s1

InChI key

VAKHFAFLRUNHLQ-PEBJKXEYSA-N

General description

A potent and competitive inhibitor of PKA (Ki = 1.7 nM).
A potent and competitive inhibitor of protein kinase A (Ki = 1.7 nM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
PKA
Product does not compete with ATP.
Reversible: no
Target Ki: 1.7 nM against protein kinase A

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-Thr-Tyr-Ala-Asp-Phe-Ile-Ala-Ser-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Glass, D.B., et al. 1989. J. Biol. Chem. 264, 8802.
Kemp, B.E., et al. 1988. Methods Enzymol.159, 173.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service