Skip to Content
MilliporeSigma
All Photos(1)

Documents

539649

Sigma-Aldrich

PKCθ/δ Inhibitor

Synonym(s):

PKCθ/δ Inhibitor, 5-(3,4-Dimethoxyphenyl)-4-(1H-indol-5-ylamino)-3-pyridinecarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18N4O2
Molecular Weight:
370.40
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

ethanol: 1 mg/mL
DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable pyridinecarbonitrile compound that acts as a potent, ATP-competitive (Ki = 79 nM, using PKCθ KD in coupled PK/LDH assays), and isozyme-selective PKCθ inhibitor (IC50 = 70 nM) with much reduced potency against other novel PKCs (IC50 = 0.35, 2.33, and 16.35 µM against δ, ε, and η isoforms, respectively) and little or no activity against the conventional PKCβ and atypical PKCζ (IC50 >50 µM) as well as a panel of 18 other commonly studied cellular kinases (IC50 >10 µM). Shown to preferentially inhibit IL-2 production from anti-CD3- and anti-CD28-activated T cells derived from wild-type over PKCθ-knockout mice (IC50 = 58 and 231 nM, respectively).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Cole, D.C., et al. 2008. J. Med. Chem.51, 5958.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service