Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

539212

Sigma-Aldrich

HMTase Inhibitor III, BIX-01338

The Protein Methyltransferase Inhibitor II, BIX-01338 controls the biological activity of Protein Methyltransferase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

HMTase Inhibitor III, BIX-01338, EHMT1/GLP Inhibitor I, EHMT2/G9a Inhibitor III, 1-(4-(4-methoxybenzoyloxy)phenethyl)-2-(4-(trifluoromethyl)benzamido)-1H-benzo[d]imidazole-5-carboxylic acid, Hydrate, 1-(2-(4-(4-Methoxybenzoyloxy)phenyl)ethyl)-2-(4-trifluoromethylbenzoylamino)-1H-benzoim

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H24F3N3O6 · xH2O
Molecular Weight:
603.54 (anhydrous basis)
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(C1=CC=C(C=C1)C(F)(F)F)NC2=NC3=C(N2CCC4=CC=C(OC(C5=CC=C(C=C5)OC)=O)C=C4)C=CC(C(O)=O)=C3.[xH2O]

General description

A cell-permable amino-benzimidazolo compound that inhibits a broad-spectrum of histone methyltransferases, including PRMT1, SET7/9, G9a, SUV39H1, and GLP (effective conc. = 15 µM) in a SAM-competitive manner. BIX-01338 is reported to affect the growth of mES, MEF, and HeLa cells at concentrations needed for HMT inhibition, limiting its use in HMTase studies to cell-free systems only.
A cell-permeable amino-benzimidazolo compound that is shown to inhibit a broad-spectrum of histone methyltransferases, including the PRMT1 H4R3 me2 activity, SET7/9 H3K4 me activity, G9a H3K9 me2 activity, as well as the H3K9 me3 activity of SUV39H1 (wild-type and H320R hyperactive mutant) and GLP (effective conc. = 15 µM) in a SAM-competitive manner. BIX-01338 is reported to affect the growth of mES, MEF, and HeLa cells at 13.3 µM, while failing to affect cellular H3K9me/me2/me3 and H3K27me2 levels at a lower non-cytotoxic concentration of 3.3 µM, limiting its use in HMTase studies to cell-free systems only.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Kubicek, S., et al. 2007. Mol. Cell25, 473.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service