Skip to Content
MilliporeSigma
All Photos(1)

Documents

539165

Sigma-Aldrich

Proteasome Inhibitor Set II

The Proteasome Inhibitor Set II controls the biological activity of Proteasome. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

ambient

storage temp.

−20°C

General description

Contains 5 mg of ALLN (Cat. No. 208719), 100 μg each of Epoxomicin (Cat. No. 324800), and clasto-Lactacystin β-Lactone (Cat. No. 426102). Supplied with a data sheet.
Proteasomes are large multi-subunit protease complexes, which selectively degrade intracellular proteins. A protein marked for degradation is attached to multiple molecules of ubiquitin, a 76-amino acid protein, which escorts it for rapid hydrolysis, by the 26S proteasome. The proteolytic core of this complex consists of the 20S proteasome, which contains multiple peptidase activities and has a molecular mass of ~700 kDa. Recently several potent, cell-permeable proteasome inhibitors have been developed.



The Calbiochem brand offers a set that includes 3 proteasome inhibitors with varying inhibtory effects.



The table below outlines some of the physical and biological properties of these compounds.

Biochem/physiol Actions

Primary Target
calpain 1, calpain 2
Secondary Target
cathepsin B, cathepsin L

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service