Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

539129

Millipore

Protease Inhibitor Cocktail VIII

liquid, for the inhibition of cysteine proteases, including calpains, cathepsins and papain. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$167.00

$167.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$167.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.28
solubility:
water: soluble

$167.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Protease Inhibitor Cocktail Set VIII, The Protease Inhibitor Cocktail Set VIII controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

solubility

water: soluble

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service