Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

538498

Sigma-Aldrich

RGS4 Inhibitor, CCG-

Synonym(s):

RGS4 Inhibitor, CCG-, 4-Butyl-2-ethyl-1,2,4-thiadiazolidine-3,5-dione, CCG203769

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O2S
Molecular Weight:
202.27
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

colorless

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable, thiadiazolidinone based, thiol reactive compound that covalently binds to the regulator of G protein signaling 4 (RGS4) and irreversibly inhibits its activity (IC50 = 17 nM). Exhibits high selectivity over RGS19 and RGS16 (IC50 = 140 nM and 6 µM, respectively) and has negligible effects towards RGS7, RGS8, and papain IC50 = 100, 70, and 100 µM, respectively). Displays ~300-fold greater selectivity over GSK-3beta (IC50 = 5.4 µM). Shown to disrupt RGS4/Gαo interaction and reverse the Gαo-induced membrane translocation of GFP-tagged RGS4 in HEK293 cells (~3 µM). Enhances Gαq-dependent Ca2+ signaling activated by the M3 muscarinic receptor. Potentiates Gαi-dependent muscarinic bradycardia and reverses D2 antagonist raclopride-induced akinesia and bradykinesia in a murine model of the movement disorder in Parkinson′s disease.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
RGS4
Reversible: no
Target IC50: 17 nM for RGS4 inhibition

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Blazer, L.L., et al. 2015. ACS Chem. Neurosci.6, 311.

Turner, E., et al. 2012. ACS Med. Chem. Lett..3, 146.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service