Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.38437

Sigma-Aldrich

MGAT2 Inhibitor, Cpd24d

Synonym(s):

MGAT2 Inhibitor, Cpd24d, 5-(N-(2,4-difluorophenyl)sulfamoyl)-7-(2-oxopyrrolidin-1-yl)-N-(4-(trifluoromethyl)phenyl)indoline-1-carboxamide , Acyl CoA:monoacylglycerol acyltransferase 2 inhibitor, Cpd24d

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H21F5N4O4S
Molecular Weight:
580.53
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, bioavailable, indoline derivative that acts as a highly potent inhibitor of acyl CoA:monoacylglycerol acyltransferase 2 inhibitor (MGAT2, IC50 = 3.4 nM). Displays good microsomal stability and desirable pharmacokinetic properties (F = 52%; AUC 0 - 8 h = 842 ng/h/ml). Effectively suppresses the elevation of plasma triacylglyceride levels after olive oil challenge in C57BL/6J mice (10 - 30 mg/kg; p.o.).
A cell-permeable, bioavailable, indoline derivative that acts as a highly potent inhibitor of acyl CoA:monoacylglycerol acyltransferase 2 inhibitor (MGAT2, IC50 = 3.4 nM). Displays good microsomal stability and desirable pharmacokinetic properties (F = 52%; AUC 0 - 8 h = 842 ng/h/ml). Effectively suppresses the elevation of plasma triacylglyceride levels after olive oil challenge in C57BL/6J mice (10 - 30 mg/kg; p.o.).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MGAT2
Reversible: yes
Target IC50: 3.4 nM for MGAT inhibition

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Sato, K., et al. 2015. J. Med. Chem.58, 3892.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service