Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.38356

Sigma-Aldrich

SRPIN803

Synonym(s):

SRPIN803, 6-(4-Hydroxy-3-methoxybenzylidene)-5-imino-2-(trifluoromethyl)-5H-[1,3,4]thiadiazolo[3,2-a]pyrimidin-7(6H)-one, SRPK1 Inhibitor II, Casein Kinase II Inhibitor XIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9F3N4O3S
Molecular Weight:
370.31
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 20 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, bioavailable, non-toxic thiadiazolopyrimidinone based compound that acts a potent, reversible and ATP-competitive dual inhibitor of CK2 & SRPK1 (IC50 = 0.2 & 2.4 µM, respectively). Displays excellent selectivity in a 306-kinase panel (% inhibition at 10 µM = 82.5, 70.9, 59.9 & 13.7 for CK2α1/β, CK2α2/β, SRPK1 & SRPK2, respectively). Synergistically suppresses VEGF production in a time and dose-dependent manner and down-regulates several angiogenesis genes expression (10 µM in ARPE-19 cells). Shown to be highly efficacious than SRPIN340 (Cat. No. 504293) and significantly inhibit choroidal neovascularization in a mouse model of age-related macular degeneration (intravitreous injection or topical administration).
A cell-permeable, bioavailable, non-toxic thiadiazolopyrimidinone based compound that acts a potent, reversible and ATP-competitive dual inhibitor of CK2 & SRPK1 (IC50 = 0.2 & 2.4 µM, respectively). Displays excellent selectivity in a 306-kinase panel (% inhibition at 10 µM = 82.5, 70.9, 59.9 & 13.7 for CK2α1/β, CK2α2/β, SRPK1 & SRPK2, respectively). Synergistically suppresses VEGF production in a time and dose-dependent manner and down-regulates several angiogenesis genes expression (10 µM in ARPE-19 cells). Shown to be highly efficacious than SRPIN340 (Cat. No. 504293) and significantly inhibit choroidal neovascularization in a mouse model of age-related macular degeneration (intravitreous injection or topical administration).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CK2, SRPK1
Reversible: yes
Target IC50: 0.2 & 2.4 &micro

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Morooka, S., et al. 2015. Mol. Pharmacol.88, 316.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service