Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.38244

Sigma-Aldrich

Cyclorasin 9A5

Synonym(s):

Cyclorasin 9A5, Cyclic Ras Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$317.00

$317.00


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
$317.00

About This Item

Empirical Formula (Hill Notation):
C75H108FN25O13
Molecular Weight:
1586.82
UNSPSC Code:
12352200
NACRES:
NA.77

$317.00


Please contact Customer Service for Availability

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

water: 100 mg/mL

storage temp.

−20°C

General description

A cell-permeable 11-mer cyclic peptide that displays high-affinity towards Ras-GTP (IC50 = 120 nM for K-Ras G12V) and inhibits Ras signaling. Shown to readily localize into cytoplasm and directly bind to Ras-GTP near the effector-binding site in a reversible manner. Dose-dependently impairs Ras-B-Raf interaction and blocks EGF-stimulated activation of Akt, MEK & ERK (IC50 = 3.1 µM in H1299 cells). Efficiently induces lung cancer cell growth arrest and triggers apoptosis (LD50 = 3.0 µM in H1299). NOTE: ala = D-Alanine; nal = D-2-naphthylalanine; nle = D-norleucine; Fpa = L-4-Fluorophenylalanine

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Ras-GTP
Reversible: yes
Target IC50: 120 nM for K-Ras G12V

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

cyclo(-Trp-Thr-ala-Arg-Arg-Arg-nal-Arg-Fpa-nle-Gln)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Upadhyaya. P., et al. 2015. Angew. Chem. Int. Ed.54, 7602.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service