Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.38184

Sigma-Aldrich

Synoviolin Inhibitor, LS-102

Synonym(s):

Synoviolin Inhibitor, LS-102, (S)-N²-(benzo[d]oxazol-6-yl)-N⁴-(1-cyclohexylethyl)-N⁶-ethyl-N⁶-(2-(ethylamino)ethyl)-1,3,5-triazine-2,4,6-triamine, E3 Ligase SYVN Inhibitor, LS-102, LS102

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36N8O
Molecular Weight:
452.60
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

potency

35 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

purple

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable triaminotriazine that selectively inhibits the autoubiquitnation of the E3 ubiquitin ligase synoviolin (SYVN1, IC50 = 35 µM), without affecting other E3 ubiquitin ligases. Abolished the SYVN-mediated negative regulation of PGC-1β via reduced ubiquitination. Shown to induce the expression of PGC-1β in wild type mouse embryonic fibroblasts (MEF), but does not affect PGC-1β expression in SYVN knockout MEFs. Increases mitochondrial biogenesis in C57BL/6J mice. Also increases the energy expenditure and causes a significant reduction in body weight and blood glucose levels in db/db mice (50 mg/kg, i.p daily) without influencing their food intake. Suppresses the proliferation of rheumatoid synovial cells (IC50 = 5.4 µM) and reduces the severity of rheumatoid arthritis in a mouse model (4 mg/kg).
A cell-permeable triaminotriazine that selectively inhibits the autoubiquitnation of the E3 ubiquitin ligase synoviolin (SYVN1, IC50 = 35 µM), without affecting other E3 ubiquitin ligases. Abolished the SYVN-mediated negative regulation of PGC-1β via reduced ubiquitination. Shown to induce the expression of PGC-1β in wild type mouse embryonic fibroblasts (MEF), but does not affect PGC-1β expression in SYVN knockout MEFs. Increases mitochondrial biogenesis in C57BL/6J mice. Also increases the energy expenditure and causes a significant reduction in body weight and blood glucose levels in db/db mice (50 mg/kg, i.p daily) without influencing their food intake. Suppresses the proliferation of rheumatoid synovial cells (IC50 = 5.4 µM) and reduces the severity of rheumatoid arthritis in a mouse model (4 mg/kg).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Synoviolin
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yagishita, N., et al. 2012. Int. J. Mol. Med.30, 1281.

Fujita, F., et al. 2015. EMBO J.34, 12192.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service