Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.38096

Sigma-Aldrich

GluN2B NMDA Receptor Antagonist, 93-31

Synonym(s):

GluN2B NMDA Receptor Antagonist, 93-31, (S)-N-(4-(3-(butyl(3,4-dichlorophenethyl)amino)-2-hydroxypropoxy)phenyl)methanesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$275.00

$275.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$275.00

About This Item

Empirical Formula (Hill Notation):
C22H30Cl2N2O4S
CAS Number:
Molecular Weight:
489.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$275.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid (sticky)

potency

0.19 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow to yellow

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A tertiary N-alkylamine that acts as a pH dependent antagonist of GluN2B-NMDA receptors. Shown to be about 10-fold more potent in ischemic tissue (pH 6.9) than in normal healthy tissue (pH 7.6) (IC50 = 190 nM and 1.8 µM, respectively). Effectively reduces the infarct volume in damaged brain tissue in a murine model of ischemic stroke (at 10 mg/kg, ED50 ≤ 1 mg/kg) without impairing coordination or motor function. Also displays binding to hERG (IC50 = 70 nM) and alpha1-adrenergic receptor (IC50 = 4.9 µM).

Biochem/physiol Actions

Primary Target
GluN2B NMDA Receptor
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yuan, H., et al. 2015. Neuron.85, 1305.
Tahirovic, Y., et al. 2008. J. Med. Chem.51, 5506.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service