Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.33998

Sigma-Aldrich

Mitotic Tracker Probe, CDy6

Synonym(s):

Mitotic Tracker Probe, CDy6, ( E)-7-(3,5-di- tert-Butyl-2-methoxystyryl)-3-(3-(2-(dimethylamino)ethylamino)-3-oxopropyl)-5,5-difluoro-9-methyl-5H-dipyrrolo[1,2-c:1ʹ,2ʹ-f][1,3,2]diazaborinin-4-ium-5-uide, Lysosomal Tracker Probe, CDy6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H47BF2N4O2
Molecular Weight:
592.57
UNSPSC Code:
12352202

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark purple

fluorescence

λex 570 nm
λem 585 nm

storage temp.

−20°C

General description

A cell-permeable, photostable, bodipy-derived fluorescent probe that can be used for real-time imaging of lysosomal dynamics in proliferating cells undergoing mitosis. Undergoes rapid re-localization from cytoplasm to nucleus during cell division, but does not interfere with cell cycle. Displays a sharp increase in intensity during mitosis and its fluorescence displays high sensitivity to acidic environment (pH 2-4). Can also be used for time-dependent whole-body in vivo imaging of tumor xenografts (CDy6 stained HeLa cells, s.c.) in BALB/c nude mice up to 21 days. Exhibits greater brightness, higher photostability, and significantly reduced toxicity when compared to LysoTracker Red, LysoTracker Green, LysoSensor Green and HOE33342. Abs max: 570 nm; Em max: 585 with a quantum yield of 0.89.
A cell-permeable, photostable, bodipy-derived fluorescent probe that can be used for real-time imaging of lysosomal dynamics in proliferating cells undergoing mitosis. Undergoes rapid re-localization from cytoplasm to nucleus during cell division, but does not interfere with cell cycle. Displays a sharp increase in intensity during mitosis and its fluorescence displays high sensitivity to acidic environment (pH 2-4). Can also be used for time-dependent whole-body in vivo imaging of tumor xenografts (CDy6 stained HeLa cells, s.c.) in BALB/c nude mice up to 21 days. Exhibits greater brightness, higher photostability, and significantly reduced toxicity when compared to LysoTracker Red, LysoTracker Green, LysoSensor Green and HOE33342. Abs max: 570 nm; Em max: 585 with a quantum yield of 0.89.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
cytoplasm
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C). Store in a dark vial, protected from light.
Use only fresh, anhydrous DMSO for reconstitution.

Other Notes

Jeong, Y.M., et al. 2015. Chem. Biol.22, 299.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service