Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.33850

Sigma-Aldrich

TAM Receptor Kinase Inhibitor, LDC1267

Synonym(s):

TAM Receptor Kinase Inhibitor, LDC1267, TAM Receptor Blocker

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$356.00

$356.00


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
$356.00

About This Item

Empirical Formula (Hill Notation):
C30H26F2N4O5
CAS Number:
Molecular Weight:
560.55
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$356.00


Please contact Customer Service for Availability

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

Fc1c(ccc(c1)NC(=O)c4n[n](cc4OCC)c5c(cc(cc5)F)C)Oc2c3c(ncc2)cc(c(c3)OC)OC

InChI

1S/C30H26F2N4O5/c1-5-40-28-16-36(23-8-6-18(31)12-17(23)2)35-29(28)30(37)34-19-7-9-25(21(32)13-19)41-24-10-11-33-22-15-27(39-4)26(38-3)14-20(22)24/h6-16H,5H2,1-4H3,(H,34,37)

InChI key

ISPBCAXOSOLFME-UHFFFAOYSA-N

General description

A cell-permeable, bioactive, non-toxic quinoline based compound that acts as a highly potent, ATP-non-competitive, tight-binding type II TAM (Tyro3, Axl, and Mer) kinase inhibitor (IC50 = 8, 29, and 5 nM, respectively for Tyro3, Axl, and Mer). Occupies both the ATP binding cleft and an adjacent hydrophobic pocket. Also displays inhibitory activity against Met, Aurora B, Lck, and Src, but only at relatively higher concentration (IC50 = 35, 36, 51, and 338 nM, respectively). Displays modest selectivity over 39 other protein kinases (at ~1 µM). Reduces the proliferation of selected cancer lines (~ 15 µM). By inhibiting TAM receptors, it enhances anti-metastatic activity of natural killer (NK) cells via TAMR/E3 ligase (Cbl-b; casitas B-lineage lymphoma-b) pathway modulation. Reduces the number and size of 4T1 micro metastases in the liver of an orthotopic murine model of metastatic breast cancer without apparently affecting the primary mammary tumor. Also shown to reduce metastatic spreading of B16F10 melanomas, an effect that is abolished upon NK1.1 depletion.
TAM Receptor Kinase Inhibitor, LDC1267, is a cell-permeable, tight-binding "type II" TAM kinase inhibitor (IC₅₀ = 8, 29 & 5 nM for Tyro3, Axl & Mer). Enhances anti-metastatic activity of NK cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Tyro3, Axl, and Mer kinases
Secondary Target
Met, Aurora B, Lck and Src kinases
Target IC50: 8, 29, and 5 nM for Tyro3, Axl, and Mer, respectively

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Paolino, M., et al. 2014. Nature.507, 508.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service