Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.33756

Sigma-Aldrich

Ac-CoA Synthase Inhibitor

Synonym(s):

Ac-CoA Synthase Inhibitor, 1-(2,3-di(Thiophen-2-yl)quinoxalin-6-yl)-3-(2-methoxyethyl)urea, N-(2,3-di-2-Thienyl-6-quinoxalinyl)-Nʹ-(2-methoxyethyl)urea, Acyl-CoA Synthetase 2 short-chain Inhibitor; short-chain Acetyl-CoA Synthetase 2 Inhibitor, ACSS2 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$317.00

$317.00


Please contact Customer Service for Availability


Select a Size

Change View
25 MG
$317.00

About This Item

Empirical Formula (Hill Notation):
C20H18N4O2S2
CAS Number:
Molecular Weight:
410.51
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$317.00


Please contact Customer Service for Availability

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[s]1c(ccc1)c2nc3c(nc2c4[s]ccc4)ccc(c3)NC(=O)NCCOC

InChI

1S/C20H18N4O2S2/c1-26-9-8-21-20(25)22-13-6-7-14-15(12-13)24-19(17-5-3-11-28-17)18(23-14)16-4-2-10-27-16/h2-7,10-12H,8-9H2,1H3,(H2,21,22,25)

InChI key

CTPVNWVUAGMHEJ-UHFFFAOYSA-N

General description

A cell-permeable quinoxaline based compound that acts as a potent, reversible inhibitor of acetyl CoA synthetase (ACSS2; IC50 ~ 600 nM in a luciferase based assay). Exhibits selectivity over ACSF2 and ACSL5 acyl-CoA synthetases. Efficiently blocks the uptake of 14C tracer carbons from acetate into lipids (IC50 = 6.8 µM) and for histone acetylation (IC50 = 5.5 µM) in HepG2 cells.

Biochem/physiol Actions

Primary Target
ACSS2
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Comerford, S.A., et al. 2014. Cell.159, 1591.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service