Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.33469

Sigma-Aldrich

GPR120 Agonist IV, cpdA

Synonym(s):

GPR120 Agonist IV, cpdA, 2-(3-(2-Chloro-5-(trifluoromethoxy)phenyl)-3-azaspiro[5.5]undecan-9-yl)acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$305.00

$305.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$305.00

About This Item

Empirical Formula (Hill Notation):
C19H23ClF3NO3
CAS Number:
Molecular Weight:
405.84
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$305.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)Oc1cc(c(cc1)Cl)N2CCC3(CC2)CCC(CC3)CC(=O)O

InChI

1S/C19H23ClF3NO3/c20-15-2-1-14(27-19(21,22)23)12-16(15)24-9-7-18(8-10-24)5-3-13(4-6-18)11-17(25)26/h1-2,12-13H,3-11H2,(H,25,26)

InChI key

WUJVPELCYCESAP-UHFFFAOYSA-N

General description

A cell-permeable azaspiroundecanyl acetic acid compound that acts as a selective Gpr120 (FFAR4) agonist toward both human and murine species (EC50 = 60 nM/IP3 production and 350 nM/β-Arrestin-2 recruitment using cells expressing human or murine Gpr120), while being ineffective toward Gpr40 and is about 100-fold more effective than docosahexaenoic acid/DHA in stimulating serum response element/SRE promoter-mediated reporter transcription in mGPR120-expressing HEK 293. Selectively suppresses (10 µM) chemotaxis toward adipocyte-conditioned medium and LPS-induced inflammatory responses in primary macrophages derived from wt, but not Gpr120-knockout, mice in vitro, including IκB degradation as well as phosphorylations of Tak1, Ikk-β, and JNK, and is efficacious in improving glucose tolerance & insulin sensitivity in vivo among wt, but not Gpr120-knockout, mice on high-fat diet (HFD) when supplemented (30 mg/kg) in the diet for the last 5 wks of a 20-wk HFD period without affecting animal body weight, resulting in decreased hyperinsulinemia, hepatic steatosis, adipose tissue macrophage (ATM) infiltration and inflammation. Unlike the Gpr40/Gpr120 dual agonist DHA, this compound does not stimulate Gpr40-mediated insulin secretion from MIN6 β cell line or ehances Glucose-stimulated insulin secretion (GSIS) from β cells in cultured islets from wt or Gpr120-knockout mice.
A cell-permeable, orally active, non-toxic, azaspiroundecanyl acetic acid based compound that acts as a highly potent, selective and reversible activator of GPR120 in murine and human cells (EC50 = 350 nM for β-arrestin 2 recruitment). Induces concentration-dependent increase in inositol-1,4,5-triphosphate (IP3) production in GRP120 expressing cells. Exhibits about 50-fold higher sensitivity for GRP120 than docosahexaenoic acid, however, it displays very poor affinity towards GRP40. Effectively reduces hepatic steatosis and improves glucose tolerance, hyperinsulinemia, and insulin sensitivity in obese mice fed high fat diet. Strongly inhibits LPS-induced phosphorylation of TAK1, IKKβ & JNK and reduces IκB degradation in macrophages.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GPR120
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Oh, D.Y., et al. 2014. Nat. Med.8, 942.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service