Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.33304

Sigma-Aldrich

DSM1

≥97% (HPLC), solid, DHODH inhibitor, Calbiochem®

Synonym(s):

DHODH Inhibitor, DSM1, 5-Methyl-N-(2-naphthyl)[1,2,4]triazolo[1,5-a]pyrimidin-7-amine, Malaria Parasite Dihydroorotate Dehydrogenase Inhibitor, P. falciparum Dihydroorotate Dehydrogenase Inhibitor, Plasmodium falciparum Dihydroorotate Dehydrogenase Inhibitor; T

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13N5
CAS Number:
Molecular Weight:
275.31
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

DHODH Inhibitor, DSM1,

Assay

≥97% (HPLC)

Quality Level

form

solid

potency

15 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

CC1=NC2=NC=NN2C(=C1)NC3=CC4=CC=CC=C4C=C3

General description

A cell-permeable triazolopyrimidine compound that potently inhibits malaria parasite Plasmodium falciparum dihydroorotate dehydrogenase- (PfDHODH-) catalyzed electron transfer from FMNH2 to Coenzyme Q10 (CoQ or ubiquinone) in a potent (IC50 = 47 nM), CoQ-competitive (Ki = 15 nM), and species-specific manner, without affecting PfDHODH-catalyzed electron transfer from DHO to FMN or the enzymatic activity of human DHODH (<5% inhibition at 100 µM). Reported to inhibit the proliferation of P. falciparum parasites (EC50 = 79 nM against 3D7 strain) in vitro, including the drug-resistant strain Dd2 (EC50 = 140 nM), while displaying no inhibitory potency against murine lymphocytic leukemia cell line L1210 even at concentrations as high as 10 µM. DSM1 is not recommended for in vivo use due to reduced plasma exposure upon repeat dosing in mice.
A cell-permeable triazolopyrimidine compound that potently inhibits malaria parasite Plasmodium falciparum dihydroorotate dehydrogenase- (PfDHODH-) catalyzed electron transfer from FMNH2 to Coenzyme Q10 (CoQ or ubiquinone) in a potent (IC50 = 47 nM), CoQ-competitive (Ki = 15 nM), and species-specific manner, without affecting PfDHODH-catalyzed electron transfer from DHO to FMN or the enzymatic activity of human DHODH (<5% inhibition at 100 µM). Reported to inhibit the proliferation of P. falciparum parasites (EC50 = 79 nM against 3D7 strain) in vitro, including the drug-resistant strain Dd2 (EC50 = 140 nM), while displaying no inhibitory potency against murine lymphocytic leukemia cell line L1210 even at concentrations as high as 10 µM. DSM1 is not recommended for in vivo use due to reduced plasma exposure upon repeat dosing in mice.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
DHODH Inhibitor, DSM1, is a highly potent, competitive, tight-binding inhibitor of Plasmodium falciparum dihydroorotate dehydrogenase (IC₅₀ = 47 nM; Ki = 15 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Pf*DHODH

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Guler, J.L., et al. 2013. PLoS Pathog.9, e1003375.

Ke, H., et al. 2011. Eukaryot. Cell.10, 1053.
Ganesan, S.M., et al. 2011. Mol. Biochem. Parasitol.177, 29.
Booker, M.L., et al. 2010. J. Biol. Chem.285, 33054.
Gujjar, R., et al. 2009. J. Med. Chem.52, 1864.
Phillips, M.A., et al. 2008. J. Med. Chem.51, 3649.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service