Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

5.33299

Sigma-Aldrich

q G Protein Inhibitor, BIM-46187

Synonym(s):

q G Protein Inhibitor, BIM-46187, (S,2R,2ʹR)-3,3ʹ-Disulfanediylbis(2-amino-1-((S)-8-(cyclohexylmethyl)-2-phenyl-5,6-dihydroimidazo[1,2-a]pyrazin-7(8H)-yl)propan-1-one) Tetrahydrochloride, 4HCl, BIM46187, G Protein Inhibitor, BIM-46187

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$331.00

$331.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$331.00

About This Item

Empirical Formula (Hill Notation):
C44H58N8O2S2 · 4HCl · xH2O
Molecular Weight:
940.96 (anhydrous basis)
UNSPSC Code:
12352202
NACRES:
NA.77

$331.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

1-3 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

water: 10 mg/mL
DMSO: 25 mg/mL

storage temp.

−20°C

General description

A cell-permeable tetrahydroimidazo[1,2a]pyrazine dimer that directly binds to Gα subunit and inhibits heterotrimeric G protein signaling. Shown to selectively abolish Gαq signaling in HEK294 and CHO cells via trapping Gαq in the empty pocket conformation by permitting GDP exit but interdicting GTP entry. In addition, elicits a dose-dependent analgesic effect in carrageenan-induced hyperalgesia (0.1-1 mg/kg, i.v.) and in a chronic constriction injury mouse model (0.3-3 mg/kg, i.v.), with synergistic effect when co-administered with morphine.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
A cell-permeable tetrahydroimidazo[1,2a]pyrazine dimer that directly binds to Gα subunit and inhibits heterotrimeric G protein signaling. Shown to selectively abolish Gαq signaling in HEK294 and CHO cells via trapping Gαq in the empty pocket conformation by permitting GDP exit but interdicting GTP entry. In addition, elicits a dose-dependent analgesic effect in carrageenan-induced hyperalgesia (0.1-1 mg/kg, i.v.) and in a chronic constriction injury mouse model (0.3-3 mg/kg, i.v.), with synergistic effect when co-administered with morphine.



Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a HCl salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Schmitz, A. et al. 1999. Chem. & Biol.21, 890.

Ayoub, M. A., et al. 2009. J. Biol. Chem.284, 29136.

Favre-Guilmard, C., et al. 2008. Eur. J. Pharm.594, 70.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service