Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

5.32626

Sigma-Aldrich

Ral Activation Inhibitor, BQU57

Synonym(s):

Ral Activation Inhibitor, BQU57, 6-Amino-1,3-dimethyl-4-(4-(trifluoromethyl)phenyl)-1,4-dihydropyrano[2,3-c]pyrazole-5-carbonitrile, Ral Inhibitor, RalA Activation Inhibitor, RalB Activation Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13F3N4O
CAS Number:
Molecular Weight:
334.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

NC(OC1=C(C(C)=NN1C)C2C3=CC=C(C=C3)C(F)(F)F)=C2C#N

General description

A cell-permeable compound that specifically, reversibly, and stoichiometrically (1:1) binds to the GDP-bound Ral A/B in an allosteric manner (Kd = 7.7 µM) and blocks its activation.
A cell-permeable dihydropyranopyrazole compound that binds and locks RalA/B in the inactive GDP-bound form by targeting an allosteric site close to the guanine nucleotide-binding pocket (Kd = 7.7 µM binding study by ITC using RalB) in a 1:1 stoichiometric ratio, while exhibiting little affinity toward free or GTP-bound Ral. Shown to inhibit anchorage-independent growth of human lung cancer cell lines H358 & H2122 in vitro (IC50 = 1.3 & 2.0 µM, respectively, in 2-4 wks by soft agar colony formation assays) by reducing cellular level of ATP-bound, active RalA/B (by >90% with 10 µM BQU57 treatment for 3 h; assessed by by RALBP1-agarose pull-down). BQU57 is bioavailable in mice via intraperitoneal injection (Plasma T1/2 = 1.5 h, AUC0-5 h = 28.6 µg·h/mL post single 50 mg/kg i.p. dosage) with overall tissue distribution, including brain (µg drug/g tissue = 4.1/liver, 2.4/kidney, 2.1/heart, 1.8/lung, 1.4/brain, 1.2/H2122 tumor; 3 h post single 50 mg/kg i.p. dosage). Shown to selectively downregulate leves of active RalA/B, but not Ras or RhoA, in H2122-derived tumor in mice in a dose-dependent manner (64% and 86% reduction of ATP-bound RalA and RalB, respectively, 3 h post single 50 mg/kg i.p. dosage) and effectively suppress H2122 tumor expansion when administered via daily i.p. dosing (by 70% 22 d post H2122 inoculation with 21 daily 50 mg/kg/d i.p. dosages) in vivo.
A cell-permeable dihydropyranopyrazole compound that binds and locks RalA/B in the inactive GDP-bound form by targeting an allosteric site close to the guanine nucleotide-binding pocket (Kd = 7.7 µM binding study by ITC using RalB) in a 1:1 stoichiometric ratio, while exhibiting little affinity toward free or GTP-bound Ral. Shown to inhibit anchorage-independent growth of human lung cancer cell lines H358 & H2122 in vitro (IC50 = 1.3 & 2.0 µM, respectively, in 2-4 wks by soft agar colony formation assays) by reducing cellular level of ATP-bound, active RalA/B (by >90% with 10 µM BQU57 treatment for 3 h; assessed by by RALBP1-agarose pull-down). BQU57 is bioavailable in mice via intraperitoneal injection (Plasma T1/2 = 1.5 h, AUC0-5 h = 28.6 µg·h/mL post single 50 mg/kg i.p. dosage) with overall tissue distribution, including brain (µg drug/g tissue = 4.1/liver, 2.4/kidney, 2.1/heart, 1.8/lung, 1.4/brain, 1.2/H2122 tumor; 3 h post single 50 mg/kg i.p. dosage). Shown to selectively downregulate leves of active RalA/B, but not Ras or RhoA, in H2122-derived tumor in mice in a dose-dependent manner (64% and 86% reduction of ATP-bound RalA and RalB, respectively, 3 h post single 50 mg/kg i.p. dosage) and effectively suppress H2122 tumor expansion when administered via daily i.p. dosing (by 70% 22 d post H2122 inoculation with 21 daily 50 mg/kg/d i.p. dosages) in vivo.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Ral-GDP

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Yan, C., et al. 2014. Nature.515, 443.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service