Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.32583

Sigma-Aldrich

TLR1/TLR2 Agonist II, CU-T12-9

Synonym(s):

TLR1/TLR2 Agonist II, CU-T12-9, N-Methyl-4-nitro-2-(4-(4-(trifluoromethyl)phenyl)-1H-imidazol-1-yl)aniline, CUT129

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13F3N4O2
CAS Number:
Molecular Weight:
362.31
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A diphenyl substituted imidazole based compound that directly and selectively targets TLR1/2 and induces their dimerization and activates TLR1 & 2 signaling leading to NF-κB and AP-1 activation (KD = 182 and 478 nM, respectively; EC50 = 52.9 nM for TLR2 in HEK-Blue cells over-expressing hTLR2). Does not affect the dimerization of TLR2/TLR6 and exhibits poor affinity towards TLR3, TLR4, TLR5, TLR7 and TLR8. Shown to compete with Pam3CSK4 (Cat. No. 506350; Ki = 45.4 nM) for binding to TLR1/2 interface and enhance heterodimerization. Up-regulates the expression of TLR1, TLR2, TNFα, IL-10, and iNOS in RAW 264.7 macrophages in a time-dependent manner. Shown to be non-toxic up to 100 µM concentration.

Please note that the molecular weight for this compound is batch-specific due to variable water content.
A non-cytotoxic (up to 100 µM) diphenyl-substituted imidazole compound that exhibits high affinity toward both TLR1 & TLR2 (KD = 182 nM & 478 nM, respectively) and induces TLR1/2 heterodimerization, effectively completing against Pam3CSK4 (Cat. No. 506350) for TLR1/2 binding (Ki = 45.4 nM; [Pam3] = 20 mg/mL; [TLR1/2] = 80 nM). Shown to potently induce secreted embryonic alkaline phosphatase (SEAP) production from human TLR2-, but not TLR3-, 4-, 5-, 7-, 8-, transfected HEK293 (EC50 = 52.9 nM) in an NF-κB inhibitor Triptolide-(Cat. No. 645900) blockable manner via selective TLR1/2, but not TLR2/6, heterodimer activation. Reported to induce comparable NF-κB-dependent reporter transcription as 100 ng/mL Pam3CSK4 when administered to human macrophage U937 cultures at 5 µM concentration (24 h) and effectively trigger NO production in both murine Raw 264.7 and primary rat macrophage cultures (ECmax = 1.2 & 0.4 µM, respectively; 24 h), blockable by TLR1/2 antagonist CU-CPT22 (Cat. No. 614305), but not TLR4 antagonist TAK-242 (Cat. No. 614316 & 508336). Likewise, both CU-T12-9 and Pam3CSK4 (1 µM & 50 ng/mL, respectively) are demonstrated to induce similar time-dependent induction of TLR1, TLR2, TNF-α, iNOS, IL-10 mRNA in Raw 264.7 cells.
TLR1/TLR2 Agonist & CU-T12-9

Biochem/physiol Actions

Primary Target
TLR1/TLR2
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cheng, K., et al. 2014. Manuscript in preparation.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service