Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.32372

Sigma-Aldrich

THZ1

≥96% (HPLC), solid, Cdk7 inhibitor, Calbiochem®

Synonym(s):

Cdk7 Inhibitor VIII, THZ1, ( E)-N-(3-(5-Chloro-4-(1H-indol-3-yl)pyrimidin-2-ylamino)phenyl)-4-(4-(dimethylamino)but-2-enamido)benzamide, 4HCl, THZ1, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H28ClN7O2 · 4HCl
Molecular Weight:
711.90
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Cdk7 Inhibitor VIII, THZ1,

Assay

≥96% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

General description

THZ1 is a highly potent, selective and irreversible inhibitor of Cdk7 (IC50 = 3.2 nM). Acts by allosterically and covalently modifying Cys312 located outside the canonical kinase domain. Shown to block the phosphorylation of Ser5 and Ser7 in RNAPII CTD, the intracellular substrate of Cdk7 (<250 nM, 4 h in Jurkat cells). Does not affect the activity of C312S mutated Cdk7. Exhibits comparatively week activity towards Cdk12 (IC50 = 250 nM), Cdk1, and Cdk2. Suppresses the proliferation and cell viability of T-ALL and CLL cell lines and induces apoptosis with significant reduction in the levels of MCL-1 and XIAP. Also induces a significant reduction in the transcript and protein levels of RUNX1, TAL, and GATA3 (~ 50 nM). Suppresses the growth of KOPTK1 cells xenografts in murine model (10 mg/kg, b.i.d).

Biochem/physiol Actions

THZ1, an acrylamide-bearing phenylaminopyrimidine that, despite its ATP-competitive affinities toward multiple kinases (IC50 ≤1 µM against ATP probe binding to 22 kinases in a KiNativ panel screening), only effectively acts as an irreversible inhibitor against Cdk7 & Cdk12, and likely also Ckd13 & Cdk19, due to conformational positioning of a C-terminal Cys outside the kinase domain (C312 of human & murine Cdk7; C1539, C1517, C349 of human Cdk12, Cdk13, Cdk19, respectively) adjacent to the drug′s acrylamide moiety in the ATP cleft, allowing covalent bond formation. Due to the irreversible mode of action, 3 to 5 h incubation is shown to most effectively inhibit intracellular Ckd7 & Cdk12 (Effective conc. 100 to 500 nM in 293A, HeLa S3, Jurkat, and Loucy cultures). Likewise, drug washout following THZ1 incubation is demonstrated not to affect its potency against cellular Cdk7 substrates phosphorylation (RNAPII S2/5/7, Cdk1 T161, Cdk2 T160, Cdk9 T186) or cell proliferation. A strong enrichment of factors involved in Cdk7 substrate RNAPII/RPB1-driven transcriptional regulation is found among THZ1-sensitive cell lines with T cell acute lymphoblastic leukemia (T-ALL; IC50 in nM = 0.49/KOPTK1, 0.54/Loucy, 0.61/DND-41, 50 nM/Jurkat by cellular ATP measurement post 72 incubation) that depend on RUNX1 transcription for proliferation being particularly sensitive to THZ1-induced apoptotic death. Intravenous injection is efficacious in suppression KOPTK1 proliferation in mice (by 91%;10 mg/3.3 mL/Kg/12 h, BID, for 29 d one week following cancer cells injection) in vivo without affecting animal body weight.

Features and Benefits

  • Cell-permeable
  • Non-toxic
  • ATP-site directed
  • Irreversible inhibitor

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service