Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.31978

Sigma-Aldrich

ATM Inhibitor, KU 60019

Synonym(s):

ATM Inhibitor, KU 60019, KU-60019, KU60019, Ataxia Telangiectasia Mutate Kinase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H33N3O5S
CAS Number:
Molecular Weight:
547.67
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

6.3 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

InChI

1S/C30H33N3O5S/c1-19-16-32(17-20(2)37-19)18-28(35)31-23-6-7-27-22(13-23)12-21-4-3-5-25(30(21)39-27)26-14-24(34)15-29(38-26)33-8-10-36-11-9-33/h3-7,13-15,19-20H,8-12,16-18H2,1-2H3,(H,31,35)/t19-,20+

InChI key

SCELLOWTHJGVIC-BGYRXZFFSA-N

General description

A cell permeable thioxanthene based compound that acts as a highly potent inhibitor of Ataxia telangiectasia mutate (ATM) kinase inhibitor (IC50 = 6.3 nM). Displays about 270 and 1600-fold greater selectivity over DNA-PK and ATR kinase (IC50 = 1.7 and >10 µM, respectively). Has only a trivial effect on 229 protein kinases when screened in a panel at 1 µM level. Shown to block Ser15 phosphorylation of p53 and basal and radiation-induced Ser473 phosphorylation of Akt in glioma cells. Blocks radiation-induced DNA damage response and radiosensitizes human glioma cells (U87 and U1242) and mouse glioma stem cells and inhibits glioma cell migration, invasion, and growth. Also radiosensitizes orthotopic glioma xenografts and significantly extends the survival of mice.
A cell permeable thioxanthene based compound that acts as a highly potent inhibitor of Ataxia telangiectasia mutate (ATM) kinase inhibitor (IC50 = 6.3 nM). Displays about 270 and 1600-fold greater selectivity over DNA-PK and ATR kinase (IC50 = 1.7 and >10 µM, respectively). Has only a trivial effect on 229 protein kinases when screened in a panel at 1 µM level. Shown to block Ser15 phosphorylation of p53 and basal and radiation-induced Ser473 phosphorylation of Akt in glioma cells. Blocks radiation-induced DNA damage response and radiosensitizes human glioma cells (U87 and U1242) and mouse glioma stem cells and inhibits glioma cell migration, invasion, and growth. Also radiosensitizes orthotopic glioma xenografts and significantly extends the survival of mice.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
ATM kinase inhibitor, KU60019, is a cell permeable, potent inhibitor of ATM kinase inhibitor (IC₅₀ = 6.3 nM). Has 270 & 1600-fold higher selectivity over DNA-PK and ATR kinase.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ATM

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Biddlestone-Thorpe, L., et al. 2013. Clin. Cancer Res.19, 3189.
Golding, S., et al. 2009. Mol. Cancer Ther.8, 2894.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service