Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.31660

Sigma-Aldrich

GCase Activator, NCGC00188758

Synonym(s):

GCase Activator, NCGC00188758, N-(4-Ethynylphenyl)-5,7-dimethylpyrazolo[1,5-a]pyrimidine-3-carboxamide, Glucocerebrosidase Activator, Glucocerebrosidase Chemical Chaperone, NCGC758

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14N4O
Molecular Weight:
290.32
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light orange

solubility

DMSO: 2.5 mg/mL

storage temp.

2-8°C

General description

A cell-permeable pyrazolopyrimidine-carboxamide compound that directly binds to glucocerebrosidase (GCase; Kd ~ 9 µM for NT-467 fluorescent labeled GCase) and enhances its activity and serves as a chaperone for its translocation to the lysosomal compartment in fibroblasts of patients with Gaucher disease (AC50 = 5.2 and 6.5 µM for wild type and mutant N370S, respectively). Also shown to be effective in increasing GCase activity in macrophages from Gaucher′s disease patients as well as in iPSC-derived macrophages (10.7 and 3.2 fold, respectively). Normalizes reactive oxygen species production and improves chemotaxis in Gaucher disease macrophages. Shown to permeate the blood-brain barrier. Exhibits desirable pharmacokinetic properties with plasma t1/2 = 19.3 h and Cmax of 1.03 µM (following a single injection of 50 mg/kg in mice).
A cell-permeable pyrazolopyrimidine-carboxamide that is shown to enhance cellular glucocerebrosidase (GCase) activity in monocyte- & iPSC-derived human macrophage cultures (hMAC & iMAC, respectively) from healthy donors as well as patients with Type I (GCase genotype N370S/N370S or N370S/c.84dupG) or Type II (GCase genotype IVS2+1G>A/L444P) Gaucher disease (GD) due to allosteric modulation of enzyme activity as well as restoration of mutant enzyme lysosomal localization, resulting in greatly enhanced lysosomal GCase substrates processing in GD cultures (%glucosylceremide reduction = 95% in IVS2+1G>A/L444P iMAC, 46.7% in N370S/c.84dupG iMAC, ≥50% in N370S/N370S hMAC & iMAC; %glucosylsphingosine reduction = 80% in IVS2+1G>A/L444P iMAC, ≥33% in N370S/N370S hMAC & iMAC; 8 µM for 6 d). NCGC00188758 treatment is also shown to reactivate ROS production upon phagocytosis of IgG-opsonized erythrocytes in both GD hMAC & iMAC cultures as well as iMAC chemotaxis toward SDF-1. Pharmacokinetic studies reveal good liver and brain exposure following single 50 mg/kg intraperitoneal injection in mice in vivo (Tmax/Cmax/T1/2 = 0.25 h/1.03 µM/19.3 h/plasma, 0.25 h/2.76 µM/18.2 h/brain, 0.25 h/17.29 µM/28.9 h) as well as mouse liver microsome stability (55% degradation in 60 min with NADPH) & Caco-2 permeability (6.0 x 10-6 cm s-1/A to B; 6.0 x 10-6 cm s-1/B to A) in vitro. Unlike the iminosugar-based chemical chaperone Isofagomine, NCGC00188758 does not inhibit GCase activity.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Glucocerebrosidase (Gcase)
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service